missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat TIMP-1 Quantikine ELISA Kit

Product Code. 18762404
Click to view available options
Quantity:
1 Plate
6 Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18762404

Brand: R&D Systems™ SRTM100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat TIMP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Rat TIMP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies rat TIMP-1 in cell culture supernates (50 ul), serum (10 ul), edta plasma (10 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 37.50 - 2400pg/mL
Assay Sensitivity 4.5pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum
Specificity Natural and Recombinant Rat TIMP-1
Sufficient For 6 x 96-well Plates
Target Species Rat
For Use With (Application) ELISA
Gene ID (Entrez) 116510
Quantity 6 Plates
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),EDTA Plasma (10uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CLGI, Collagenase inhibitor, collagenase inhibitor), EPATIMP-1, EPO, erythroid potentiating activity, Erythroid-potentiating activity, Fibroblast collagenase inhibitor, FLJ90373, HCI, metalloproteinase inhibitor 1, TIMP metallopeptidase inhibitor 1, TIMP1, TIMPtissue inhibitor of metalloproteinase 1 (erythroid potentiating activity, Tissue inhibitor of metalloproteinases 1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.