missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Rat TNF-alpha Quantikine ELISA Kit
Click to view available options
Quantity:
6 Plates
Unit Size:
1 set
Description
The Rat TNF-alpha Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies rat TNF-alpha.
Specifications
Specifications
| Assay Range | 12.50 to 800 pg/mL |
| Assay Sensitivity | 5 pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA), Plasma (EDTA, Heparin), Plasma (Heparin), Serum, Serum |
| Specificity | Natural and Recombinant Rat TNF-alpha |
| Sufficient For | 6 x 96-well Plates |
| Target Species | Rat |
| For Use With (Application) | ELISA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction