missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ RCM 1
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
RCM 1 is a forkhead box M1 (FOXM1) transcription factor inhibitor (IC50 = 0.72 μM). Inhibits expression of FOXM1 target genes. Induces FOXM1 protein degradation in vitro. Reduces airway resistance, increases lung compliance and decreases proinflammatory cytokine production in allergen-exposed mice.
Specifications
Specifications
| Quantity | 10 mg |
| Molecular Formula | C20H12N2OS4 |
| Formula Weight | 424.58 |
| Product Type | Inhibitor |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction