Learn More
Lenz Laborglasintrumente Reaction Vessels Cylindrical with Thermostatic and Vacuum Jacket and Withdrawal Valve
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 06061482
Additional Details : Weight : 0.01000kg
Description
- With graduation
- With laboratory flange LF, with groove
- With two connectors (with ball flanges KF 15 /KF 25)
- Made of DURAN™ tubing
- Other sizes on request
Specifications
5 L | |
570 mm | |
250 mm |
Duran Glass | |
Connector KF 15 |