Learn More
Hanna Instruments™ Chromium IV Reagent Sets, For Colorimetric Water Testing
Allows users to achieve fast and accurate colorimetric measurements. Hanna Instruments™ Chromium IV Reagent Sets, For Colorimetric Water Testing are high-quality, pre-measured solutions for use with compatible benchtop and/or portable Hanna meters.
Brand: Hanna Instruments™ HI9374901
Additional Details : CAS : 7790-62-7 Weight : 0.80000kg
Description
These high quality reagents are designed to follow an adaptation of the ASTM Manual of Water and Environmental Technology, D1687-92, diphenylcarbohydrazide method for colorimetric determination of chromium VI. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the color that is produced.
- Reagent sets available for low-range and high range determinations
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
D1687-92 Diphenylcarbohydrazide | |
Compatible Hanna benchtop and portable photometers (HI83099, HI83200, HI83205, HI83206, HI96749) | |
100 | |
0 to 300μg/L chromium VI | |
Water Testing Kit |
Chromium VI low range determination | |
100 individual packets | |
1 Set | |
Cholorimetric determination |
Safety and Handling
- X100 TEST CHROMIUM VI LR
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P404-Store in a closed container.
Supplemental information
- MIXTURE LIST-Contains : potassium sulphate,sulphamic acid