missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hanna Instruments™ Iodine Reagent Sets, For Colorimetric Water Testing
Click to view available options
Quantity:
1 Set
Unit Size:
Pack of 100
Description
These high quality reagents are designed to follow an adaptation of the Standard Methods for the Examination of Water and Wastewater, 20th edition, DPD method for colorimetric determination of iodine. By simply adding the reagents to the sample as directed, a reaction will take place and a compatible meter can be used to determine the concentration from the pink color that is produced.
- Pre-made, pre-measured water testing reagents for ease of use
- Prepared with high purity chemicals manufactured in Hanna's state-of-the-art facility
- Clearly marked with expiration date and lot number on each packet and bottle for traceability
Specifications
Specifications
| For Use With (Application) | Iodine determination |
| For Use With (Equipment) | Compatible Hanna benchtop and portable photometers |
| Includes | 100 individual packets |
| No. of Tests | 100 |
| Quantity | 1 Set |
| Range | 0.00 to 12.50mg/L iodine |
| Test Method | Standard Methods, 20th edition, DPD |
| Type | Water Testing Kit |
Safety and Handling
Product Identifier
- Reagenzien für 100 Tests Jod
- Danger
- Acute toxicity Category 4
- Specific target organ toxicity Category 1
- H332-Harmful if inhaled.
- H372-Causes damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Potassium iodide, EDTA disodium salt, N,N-Diethyl-1,4-phenylenediammonium sulphate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction