Learn More
Macherey-Nagel™ Receiver Plates
Product Code. 15269992
Click to view available options
For Use With (Application):
Centrifugation and Vacuum Use
Gravity Flow, Centrifugation, Vacuum Use
Includes:
Filter Frits
Hydrophilised Filter Frits
Total Volume (Metric) Well:
10 μL
35 μL
Unit Size:
Pack of 4
Product Code. 15269992
Brand: Macherey-Nagel™ 740989.4
Macherey-Nagel 96-well receiver plates with inserted filter frits of 35 μm pore size are suitable for centrifugation and vacuum applications for high throughput work.
  • 96-well plates
  • For centrifugation and vacuum
  • 35 μm filter frits
Specifications
No. of Wells 96
Total Volume (Metric) Well 10 μL
For Use With (Application) Centrifugation and Vacuum Use
Includes Filter Frits