Learn More
Lenz Laborglasinstrumente™ Receiving Flask with PUR Coating
Click to view available options
Volume (Metric):
500 mL
1000 mL
Unit Size:
Each
Description
- With spherical joint S 35
- With highly transparent PUR coating
- Autoclavable
- Available in a range of capacities
Specifications
Specifications
| Material | Duran Glass |
| Type | Receiving Flask |
| For Use With (Equipment) | Rotary Evaporators |
| Material (Coating) | PUR |
| Volume (Metric) | 500 mL |
| Diameter (Metric) | 105 mm |