missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant B. anthracis Thymidylate kinase Protein
A DNA sequence encoding the Bacillus anthracis (strain CDC 684 / NRRL 3495) Thymidylate kinase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5954-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
WP_000677237.1 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
tmk | |
Recombinant Protein | |
E. coli | |
MKGLFVTIEG PEGSGKTTLI QSLLPYFEQK EQKVMATREP GGIAISEDIR TILHKQEYTM MEARTEALLY AAARRQHLVE KVMPALNEDY LVLCDRFIDS SLAYQGYARG LGMDKVFEIN RFATEDCMPS LTIYLDIEPE VGLARIAKDA GREVNRLDME DISFHKRVRE GYLQVVERFS DRIVLVNADQ PMEKLIEEVI QVIEDKLL |
Thymidylate kinase Protein | |
500 μg | |
Not determined. | |
Full Length | |
B. anthracis | |
His | |
Tris-based buffer, 50% glycerol |