missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine APOH Protein

Product Code. 15925998
Click to view available options
Quantity:
1 mg
10 μg
50 μg
Unit Size:
10µg
1mg
50µg
This item is not returnable. View return policy

Product Code. 15925998

Brand: enQuireBio™ QP110531mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the APOH was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 281006
Name APOH Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol APOH
Product Type Recombinant Protein
Cross Reactivity Bovine
Species Bovine
Protein Tag Untagged
Sequence MPPPALVLLL GFLCHVAIAG RTCPKPDELP FSTVVPLKRT YEPGEQIVFS CQPGYVSRGG IRRFTCPLTG LWPINTLKCM PRVCPFAGIL ENGTVRYTTF EYPNTISFSC HTGFYLKGAS SAKCTEEGKW SPDLPVCAPI TCPPPPIPKF ASLSVYKPLA GNNSFYGSKA VFKCLPHHAM FGNDTVTCTE HGNWTQLPEC REVRCPFPSR PDNGFVNHPA NPVLYYKDTA TFGCHETYSL DGPEEVECSK FGNWSAQPSC KASCKLSIKR ATVIYEGERV AIQNKFKNGM LHGQKVSFFC KHKEKKCSYT EDAQCIDGTI EIPKCFKEHS SLAFWKTDAS DVKPC
Buffer APOH protein solution is supplied in 16mM Tris-HCl pH 8.0, 280mM NaCl, 0.16mM AEBSF and 20% glycerol.
Purity or Quality Grade Greater than 70% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.