missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine ATF Protein

Product Code. 15926128
Click to view available options
Quantity:
50 mg
150 mg
1 g
Unit Size:
150mg
1g
50mg
This item is not returnable. View return policy

Product Code. 15926128

Brand: enQuireBio™ QP1109550mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Gene ID (Entrez) 280705
Name ATF Protein
Quantity 50 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol TF
Product Type Recombinant Protein
Cross Reactivity Bovine
Protein Tag Untagged
Sequence MRPAVRALLA CAVLGLCLAD PERTVRWCTI STHEANKCAS FRENVLRILE SGPFVSCVKK TSHMDCIKAI SNNEADAVTL DGGLVYEAGL KPNNLKPVVA EFHGTKDNPQ THYYAVAVVK KDTDFKLNEL RGKKSCHTGL GRSAGWNIPM AKLYKELPDP QESIQRAAAN FFSASCVPCA DQSSFPKLCQ LCAGKGTDKC ACSNHEPYFG YSGAFKCLME GAGDVAFVKH STVFDNLPNP EDRKNYELLC GDNTRKSVDD YQECYLAMVP SHAVVARTVG GKEDVIWELL NHAQEHFGKD KPDNFQLFQS PHGKDLLFKD SADGFLKIPS KMDFELYLGY EYVTALQNLR ESKPPDSSKD ECMVKWCAIG HQERTKCDRW SGFSGGAIEC ETAENTEECI AKIMKGEADA MSLDGGYLYI AGKCGLVPVL AENYKTEGES CKNTPEKGYL AVAVVKTSDA NINWNNLKDK KSCHTAVDRT AGWNIPMGLL YSKINNCKFD EFFSAGCAPG SPRNSSLCAL CIGSEKGTGK ECVPNSNERY YGYTGAFRCL VEKGDVAFVK DQTVIQNTDG NNNEAWAKNL KKENFEVLCK DGTRKPVTDA ENCHLARGPN HAVVSRKDKA TCVEKILNKQ QDDFGKSVTD CTSNFCLFQS NSKDLLFRDD TKCLASIAKK TYDSYLGDDY VRAMTNLRQC STSKLLEACT FHKP
Buffer The protein (1 mg/ml) was lyophilized with no additives.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.