missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine ECGS Protein
A cDNA sequence encoding the ECGS was constructed and used to recombinantly synthesize the protein.
137.00€ - 1060.00€
Specifications
Name | ECGS Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Bovine |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15988188
|
enQuireBio™
QP11736-5MG |
5 mg |
137.00€
5mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978188
|
enQuireBio™
QP11736-20MG |
20 mg |
212.00€
20mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968188
|
enQuireBio™
QP11736-100MG |
100 mg |
1060.00€
100mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
ECGS Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged |
Research Use Only | |
Recombinant Protein | |
Bovine | |
The Endothelial-Mitogen was lyophilized from a concentrated solution containing 0.1M NaCl and 5mM NaH2PO4, pH 7.4. |