missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine FGF 21 Protein

A cDNA sequence encoding the FGF 21 was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Name FGF 21 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Bovine
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15928648
View Documents Promotion Details
enQuireBio™
QP11870-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15908648
View Documents Promotion Details
enQuireBio™
QP11870-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15918648
View Documents Promotion Details
enQuireBio™
QP11870-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

FGF 21 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Bovine
Untagged
Greater than 98.0% as determined by:(a) Analysis by Gel Filtration. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (0.8 mg/ml) solution with 0.4 mg/ml of NaHCO3, pH 8.
Videos
SDS
Documents

Documents

Product Certifications