missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Placental Lactogen Protein
A cDNA sequence encoding the Placental Lactogen was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10826-200ug
Additional Details : Weight : 0.01000kg
Specifications
Placental Lactogen Protein | |
Research Use Only | |
Placental Lactogen Bovine is biologically active as evidenced by inducing proliferation of Nb2 cells. | |
Bovine | |
Untagged | |
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02-0.03% NaHCO3. |
200 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPASSHRGH QWICDLVRGS CLLLLLVVSN LLLCQGVEDY APYCKNQPGN CRIPLQSLFE RATLVASNNY RLAREMFNEF NKQFGEGKNF TSKVINSCHT EFMTTPNNKE AAANTEDEAL LRLVISLLHS WDEPLHQAVT ELLHRNGASP DILARAKEIE DKTKVLLEGV EMIQKRVHPG EKKNEPYPVW SEKSSLTADD EDVRQTAFYR MFHCLHRDSS KISTYINLLK CRFTPC | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |