missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Placental Lactogen Protein

Product Code. 15925288
Click to view available options
Quantity:
1 mg
200 μg
50 μg
Unit Size:
1mg
200µg
50µg
This item is not returnable. View return policy

Product Code. 15925288

Brand: enQuireBio™ QP108261mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Placental Lactogen was constructed and used to recombinantly synthesize the protein.

Specifications

Name Placental Lactogen Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity Placental Lactogen Bovine is biologically active as evidenced by inducing proliferation of Nb2 cells.
Product Type Recombinant Protein
Cross Reactivity Bovine
Species E. coli
Protein Tag Untagged
Sequence MAPASSHRGH QWICDLVRGS CLLLLLVVSN LLLCQGVEDY APYCKNQPGN CRIPLQSLFE RATLVASNNY RLAREMFNEF NKQFGEGKNF TSKVINSCHT EFMTTPNNKE AAANTEDEAL LRLVISLLHS WDEPLHQAVT ELLHRNGASP DILARAKEIE DKTKVLLEGV EMIQKRVHPG EKKNEPYPVW SEKSSLTADD EDVRQTAFYR MFHCLHRDSS KISTYINLLK CRFTPC
Buffer The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.02-0.03% NaHCO3.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.