missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken Collagen alpha-1(XII) chain Protein

A DNA sequence encoding the Gallus gallus (Chicken) Collagen alpha-1(XII) chain, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5851-ye-10ug

Additional Details : Poids : 0.01000kg

 View more versions of this product

Product Code. 15958229

Quantity Price
1 352.00€ / 10µg
6 328.49€ / 10µg
Expédition estimée
02-11-2020
Add to basket


Specifications

Specifications

Collagen alpha-1(XII) chain Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Chicken
His
Tris-based buffer, 50% glycerol
Greater than 90% as determined by SDS-PAGE.
Research Use Only
COL12A1
Recombinant Protein
Yeast
DLVFLVDGSW SVGRNNFRYI LDFMVALVSA FDIGEEKTRV GVVQYSSDTR TEFNLNQYFR RSDLLDAIKR IPYKGGNTMT GEAIDYLVKN TFTESAGARK GFPKVAIVIT DGKAQDEVEI PARELRNIGV EVFSLGIKAA DAKELKLIAS QPSLKHVFNV ANFDGIVDIQ NEI
Videos
SDS
Documents