missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chikungunya Virus E1 Envelope Protein

Product Code. p-7148272
Click to view available options
Quantity:
100 μg
500 μg
Unit Size:
100µg
500µg
This item is not returnable. View return policy

Product Code. 15923988

Brand: enQuireBio™ QP10474100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Name E1 Envelope Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Untested
Product Type Recombinant Protein
Cross Reactivity Chikungunya virus (Strain: Senegal/37997)
Species Human
Protein Tag Native
Buffer PBS, pH7.4.
Purity or Quality Grade >90% as determined by SDS-PAGE.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.