missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chinese Hamster Cathepsin D / CTSD Protein
A DNA sequence encoding the Cricetulus griseus (Chinese hamster) (Cricetulus barabensis griseus) Cathepsin D / CTSD, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5886-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
XP_003510282.1 | |
Cathepsin D / CTSD Protein | |
Research Use Only | |
CTSD | |
Recombinant Protein | |
E. coli | |
GPVSELLKNY LDAQYYGEIG IGTPPQCFTV VFDTGSSNLW VPSIHCKLLD IACWIHHKYN SGKSSTFVKN GTSFDIHYGS GSLSGYLSQD TVSVPCKSEQ PGGLKVEKQI FGEAIKQPGI TFIAAKFDGI LGMGYPSISV NNVVPVFDNL MQQKLVEKNI FSFFLNRDPT GQPGGELMLG GIDSKYYEGE LSYLNVTRKA YWQVHMDQLD VANGLTLCKG GCEAIVDTGT SLLVGPVDEV KELQKAIGAV PLIQGEYMIP CEKVSSLPSV TLKLGGKDYE LSPSKYVLKV SQGGKTICLS GFMGMDIPPP SGPLWILGDV FIGTYYTVFD RDNNRVGFAK AATL | |
Greater than 90% as determined by SDS-PAGE. |
100766628 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Chinese Hamster | |
His-SUMO | |
Tris-based buffer, 50% glycerol |