missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Monkey CD39/ENTPD1 His Protein
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
Description
Measured by its ability to hydrolyze the 5’-phosphate groups from the substrate adenosine-5’-triphosphate (ATP). The orthophosphate product is measured by a Malachite Green Phosphate Detection Kit (Catalog # DY996 ). The specific activity is >15,000 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | EHH64904.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2 and Glycerol. |
| Gene ID (Entrez) | 953 |
| Molecular Weight (g/mol) | M.W.-observed: 60-77 kDa, under reducing conditions, M.W.-theroretical: 51 kDa |
| Quantity | 20 μg |
| Source | Mouse myeloma cell line, NS0-derived cynomolgus monkey CD39/ENTPD1 protein Thr45-Val485, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | ADPase, ATPDase, ATP-diphosphatase, CD39, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, ENTPD1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPD1, NTPDase 1, NTPDase-1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction