missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus Monkey GITR/TNFRSF18 Fc Chimera

Product Code. 18725807
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18725807

Brand: R&D Systems™ 9428GR100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Cynomolgus Monkey GITR/TNFRSF18 Fc Chimera is immobilized at 0.5 μg/mL, 100 μL/well, the concentration of biotinylated recombinant human GITR Ligand/TNFSF18 that produces 50% of the optimal binding response is 8-40 ng/mL.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number XP_005545180
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 8784
Molecular Weight (g/mol) M.W. (Observed): 49-60 kDa, reducing conditions; M.W. (theoretical): 41 kDa
Quantity 100 μg
Source Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey GITR/TNFRSF18 protein Cynomolgus Monkey GITR/TNFRSF18 (Gln20-Glu155) (N-terminus) Accession # XP_005545180 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Activation-inducible TNFR family receptor, AITR
Biological Activity 8 to 40ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Cynomolgus Monkey
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein GITR/TNFRSF18
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.