missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue NS1 n Protein

A cDNA sequence encoding the Dengue NS1 n was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11643-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927898

  • 832.00€ / 500µg

Estimated Shipment: 25-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Dengue NS1 n Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
GST
Protein is >90% pure as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
100mM Tris-Hcl pH 8.0, 60mM NaCl, and 5% Glycerol.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions