missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 2 NS1 Protein
Recombinant Protein
402.00€ - 1415.00€
Specifications
Name | Serotype 2 NS1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Untested |
Product Type | Recombinant Protein |
Cross Reactivity | Dengue virus (Strain: Thailand/16681/1984) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15913998
|
enQuireBio™
QP10478-100UG |
100 μg |
402.00€
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923998
|
enQuireBio™
QP10478-500UG |
500 μg |
1415.00€
500µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The NS1 non-structural protein, is an immunogenic protein, but poses no health risks and is therefore ideal for antibody production.Specifications
Serotype 2 NS1 Protein | |
Untested | |
Dengue virus (Strain: Thailand/16681/1984) | |
His | |
>95% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
Human | |
DPBS pH 7.4. |