missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Transketolase 1 Protein

Product Code. p-7153942
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16027710

Brand: enQuireBio™ QP7207ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) Transketolase 1, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_000098614.1
Gene ID (Entrez) 947420
Name Transketolase 1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol tktA
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His-SUMO
Sequence MSSRKELANA IRALSMDAVQ KAKSGHPGAP MGMADIAEVL WRDFLKHNPQ NPSWADRDRF VLSNGHGSML IYSLLHLTGY DLPMEELKNF RQLHSKTPGH PEVGYTAGVE TTTGPLGQGI ANAVGMAIAE KTLAAQFNRP GHDIVDHYTY AFMGDGCMME GISHEVCSLA GTLKLGKLIA FYDDNGISID GHVEGWFTDD TAMRFEAYGW HVIRDIDGHD AASIKRAVEE ARAVTDKPSL LMCKTIIGFG SPNKAGTHDS HGAPLGDAEI ALTREQLGWK YAPFEIPSEI YAQWDAKEAG QAKESAWNEK FAAYAKAYPQ EAAEFTRRMK GEMPSDFDAK AKEFIAKLQA NPAKIASRKA SQNAIEAFGP LLPEFLGGSA DLAPSNLTLW SGSKAINEDA AGNYIHYGVR EFGMTAIANG ISLHGGFLPY TSTFLMFVEY ARNAVRMAAL MKQRQVMVYT HDSIGLGEDG PTHQPVEQVA SLRVTPNMST WRPCDQVESA VAWKYGVERQ DGPTALILSR QNLAQQERTE EQLANIARGG YVLKDCAGQP ELIFIATGSE VELAVAAYEK LTAEGVKARV VSMPSTDAFD KQDAAYRESV LPKAVTARVA VEAGIADYWY KYVGLNGAIV GMTTFGESAP AELLFEEFGF TVDNVVAKAK ELL
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.