missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Equine Leptin Protein
Click to view available options
Quantity:
1 mg
100 μg
20 μg
Unit Size:
100µg
1mg
20µg
Specifications
Specifications
| Name | Leptin Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | Biological active as evidenced by inducing proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Equine |
| Species | E. coli |
| Protein Tag | Untagged |
| Sequence | LWLWPYLFFI EAVPIRKVQD DTKTLIKTIV TRINDISHTQ SVSSKQRVTG LDFIPGLHPV LSLSKMDQTL AIYQQILTSL PSRNVIQISN DLENLRDLLH LLASSKSCPL PQARGLETLA SLGGVLEASL LLHRGGSPEQ AAGVS |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction