missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis A HAV P2C Protein
A cDNA sequence encoding the HAV P2C was constructed and used to recombinantly synthesize the protein.
238.00€ - 1515.00€
Specifications
Name | Hepatitis A HAV P2C Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis A |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979388
|
enQuireBio™
QP12107-100UG |
100 μg |
238.00€
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999388
|
enQuireBio™
QP12107-500UG |
500 μg |
910.00€
500µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989388
|
enQuireBio™
QP12107-1MG |
1 mg |
1515.00€
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis A HAV P2C Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis A | |
Untagged | |
Protein is >90% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM CBB, pH9.6, 0.1% SDS and 50% glycerol. |