missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg ayw Protein
A cDNA sequence encoding the HBsAg ayw was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12127-HIS-50ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis B HBsAg ayw Protein | |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
His |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
S. cerevisiae | |
HBsAg solution (1 mg/ml) containing 7.5mM phosphate buffer pH 7.2, 75mM NaCl and 50% glycerol. |