missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis B HBsAg preS1 Protein
A cDNA sequence encoding the HBsAg preS1 was constructed and used to recombinantly synthesize the protein.
137.00€ - 2815.00€
Specifications
Name | Hepatitis B HBsAg preS1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Hepatitis B |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15969458
|
enQuireBio™
QP12128-10UG |
10 μg |
137.00€
10µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989458
|
enQuireBio™
QP12128-50UG |
50 μg |
212.00€
50µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979458
|
enQuireBio™
QP12128-1MG |
1 mg |
2815.00€
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Hepatitis B HBsAg preS1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis B | |
Untagged | |
HBsAg protein was lyophilized from 0.2?m filtered (1 mg/ml) solution in 20mM PBS, pH 7.4, and 50mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGGWSSKPRQGMGTNLSVPNPLGFFPDHQLDPAFGANSNNPDWDFNPNKDHWPEAHQVGAGAFGPGFTPPHGGLLGWSPQAQGILTTVPVAPPPASTNRQSGRQPTPISPPLRDSHPQA | |
HBsAg Protein is >95% pure as determined by 10% PAGE (coomassie staining). |