missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV 4th Generation Protein

A cDNA sequence encoding the HCV 4th Generation was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12136-EC-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929488

  • 832.00€ / 500µg

Estimated Shipment: 28-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Hepatitis C HCV 4th Generation Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
Phosphate saline buffer with 25mM arginine.
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions