missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-6 Protein

A cDNA sequence encoding the HCV NS5 Genotype-6 was constructed and used to recombinantly synthesize the protein.

238.00€ - 1515.00€

Specifications

Name Hepatitis C HCV NS5 Genotype-6 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Hepatitis C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15909668
View Documents Promotion Details
enQuireBio™
QP12197-100UG
100 μg
238.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15929668
View Documents Promotion Details
enQuireBio™
QP12197-500UG
500 μg
910.00€
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15919668
View Documents Promotion Details
enQuireBio™
QP12197-1MG
1 mg
1515.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Hepatitis C HCV NS5 Genotype-6 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
Untagged
HCV NS5 Genotype-6 protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
25mM Tris-HCl, 1mM EDTA, 1.5M urea and 50% glycerol.
Videos
SDS
Documents

Documents

Product Certifications