missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 gp120 LAV Protein
A cDNA sequence encoding the HIV-1 gp120 LAV was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12245-2ug
Additional Details : Weight : 0.01000kg
Specifications
HIV-1 gp120 LAV Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
The protein solution contains 10mM Tris-Cl pH 7.6, 150mM NaCl and 0.01% Triton N-101. |
Greater than 90.0% as determined by HPLC analysis and SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
Insect | |
IPGEKLWVTV YYGVPVWKEA TTTLFCASDA KAYDTEVHNV ATHACVPTDP NPQEVVLVNV TENFNMWKND MVEQMHEDII SLWDQSLKPC VKLTPLCVSL KCTDLKNDTN TNSSSGRMIM EKGEIKNCSF NISTSIRDKV QKEYAFFYKL DIVPIDNTSY RLISCNTSVI TQACPKVSFE PIPIHYCAPA GFAILKCNNK TFNGTGPCTN VSTVQCTHGI RPVVSTQLLL NGSLAEEDVV IRSANFTDNA KTIIVQLNTS VEINCTRPNN NTRKSIRIQR GPGRAFVTIG KIGNMRQAHC NISRAKWNAT LKQIASKLRE QFGNNKTIIF KQSSGGDPEI VTHSFNCGGE FFYCNSTQLF NSTWFNSTWS TEGSNNTEGS DTITLPCRIK QFINMWQEVG KAMYAPPISG QIRCSSNITG LLLTRDGGNN NNGSEIFRPG GGDMRDNWRS ELYKYKVVKI EPLGVAPTKA KRRVVQREKR |