missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 gp120 Nef Mosaic Protein

A cDNA sequence encoding the HIV-1 gp120 Nef Mosaic was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12247-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979818

  • 1815.00€ / 1mg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV-1 gp120 Nef Mosaic Protein
Research Use Only
Recombinant Protein
E. coli
10mM Tris-HCl, pH 4.5, 25mM Sodium Phosphate and 8M urea.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
His
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers