missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 gp41 Long Protein

A cDNA sequence encoding the HIV-1 gp41 Long was constructed and used to recombinantly synthesize the protein.

238.00€ - 1815.00€

Specifications

Name HIV-1 gp41 Long Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity HIV
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15959828
View Documents Promotion Details
enQuireBio™
QP12250-EC-100UG
100 μg
238.00€
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15979828
View Documents Promotion Details
enQuireBio™
QP12250-EC-500UG
500 μg
910.00€
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15969828
View Documents Promotion Details
enQuireBio™
QP12250-EC-1MG
1 mg
1815.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

HIV-1 gp41 Long Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
8M Urea, 20mM Tris-HCl pH 8.0, 10mM b-mercaptoethanol.
Research Use Only
Recombinant Protein
E. coli
IEFPGIFRPGGGDMRDNWRSELYKYKVVKIEPLGVAPTKAKRRVVQ REKRAVGIGALFLGFLGAAGSTMGAASMTLTVQARQLLSGIVQQQNNLLR AIEAQQHLLQLTVWGIKQLQARILAVERYLKDQQLLGIWGCSGKLICTTAVPWNASWSN KSLEQIWNNMTWMEWDREINNYTSLIHSLIEESQNQQEKNEQELLELDKWASLWNWFNITNWLWYIKLFIMIVGGLVGLRIVFAVLSVVNRVRQGYSPLSFQTHLPIPRGPDRPEGIEEEGGERDRDRSIRLVNGSLALIWDDLRSLCLFSYHRLRDLLLIVTRIVELLGRRGWEALKYWWNLLQYWSQELKNSAVSLLNATAIAVAEGTDRVIEVVQGAYRAIRHIPRRIRQGLERILL
Greater than 95.0% as determined by HPLC analysis & SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications