missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 nef, Clade B Protein

A cDNA sequence encoding the HIV-1 nef, Clade B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12254-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15979838

Quantity Price
1 171.64€ / 10µg
10 161.27€ / 10µg
Estimated Shipment
30-03-2021
Add to basket


Description

Description

Specifications

Specifications

HIV-1 nef, Clade B Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Greater than 90.0% as determined by HPLC analysis and SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
Lyophilized with 10% glycerol.
Videos
SDS
Documents