missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 TAT Clade-D Protein
A cDNA sequence encoding the HIV-1 TAT Clade-D was constructed and used to recombinantly synthesize the protein.
137.00€ - 1665.00€
Specifications
Name | HIV-1 TAT Clade-D Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15929878
|
enQuireBio™
QP12265-2UG |
2 μg |
137.00€
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15919878
|
enQuireBio™
QP12265-10UG |
10 μg |
212.00€
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909878
|
enQuireBio™
QP12265-100UG |
100 μg |
1665.00€
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV-1 TAT Clade-D Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized with 10% glycerol. |