missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV HTLV-1 mosaic Protein
A cDNA sequence encoding the HTLV-1 mosaic was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12356-500ug
Additional Details : Poids : 0.01000kg
Specifications
HIV HTLV-1 mosaic Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
60mM NaCl, 50mM Tris-HCl pH 8.0, 0.25% Sarkosil, 50% glycerol and 10mM glutathione. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
HTLV-1 Mosaic protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |