missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HSV-2 gG Protein
A cDNA sequence encoding the HSV-2 gG was constructed and used to recombinantly synthesize the protein.
238.00€ - 1515.00€
Specifications
Name | HSV HSV-2 gG Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HSV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15920129
|
enQuireBio™
QP12346-100UG |
100 μg |
238.00€
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940129
|
enQuireBio™
QP12346-500UG |
500 μg |
910.00€
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15930129
|
enQuireBio™
QP12346-1MG |
1 mg |
1515.00€
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HSV HSV-2 gG Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
Untagged | |
Protein is >99% pure as determined by Analytical HPLC and Mass Spec. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HSV-2 gG solution contains 25mM Tris-HCl pH 8.0. |