missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AAGAB Protein
A cDNA sequence encoding the AAGAB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10924-20ug
Additional Details : Weight : 0.01000kg
Specifications
AAGAB Protein | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
AAGAB protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAAGVPCALV TSCSSVFSGD QLVQHILGTE DLIVEVTSND AVRFYPWTID NKYYSADINL CVVPNKFLVT AEIAESVQAF VVYFDSTQKS GLDSVSSWLP LAKAWLPEVM ILVCDRVSED GINRQKAQEW CIKHGFELVE LSPEELPEED DDFPESTGVK RIVQALNANV WSNVVMKNDR NQGFSLLNSL TGTNHSIGSA DPCHPEQPHL PAADSTESLS DHRGGASNTT DAQVDSIVDP MLDLDIQELA SLTTGGGDVE NFERLFSKLK EMKDKAATLP HEQRKVHAEK VAKAFWMAIG GDRDEIEGLS SDEEH |