missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACADSB Protein

A cDNA sequence encoding the ACADSB was constructed and used to recombinantly synthesize the protein.

137.00€ - 7875.00€

Specifications

Gene ID (Entrez) 36
Name ACADSB Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol ACADSB
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15945618
View Documents Promotion Details
enQuireBio™
QP10934-2UG
2 μg
137.00€
2µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15925618
View Documents Promotion Details
enQuireBio™
QP10934-10UG
10 μg
212.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15935618
View Documents Promotion Details
enQuireBio™
QP10934-1MG
1 mg
7875.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

36
Research Use Only
ACADSB
Human
His
The ACADSB solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT.
ACADSB Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMKSSQS EALLNITNNG IHFAPLQTFT DEEMMIKSSV KKFAQEQIAP LVSTMDENSK MEKSVIQGLF QQGLMGIEVD PEYGGTGASF LSTVLVIEEL AKVDASVAVF CEIQNTLINT LIRKHGTEEQ KATYLPQLTT EKVGSFCLSE AGAGSDSFAL KTRADKEGDY YVLNGSKMWI SSAEHAGLFL VMANVDPTIG YKGITSFLVD RDTPGLHIGK PENKLGLRAS STCPLTFENV KVPEANILGQ IGHGYKYAIG SLNEGRIGIA AQMLGLAQGC FDYTIPYIKE RIQFGKRLFD FQGLQHQVAH VATQLEAARL LTYNAARLLE AGKPFIKEAS MAKYYASEIA GQTTSKCIEW MGGVGYTKDY PVEKYFRDAK IGTIYEGASN IQLNTIAKHI DAEY
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications