missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACAT2 Protein
A cDNA sequence encoding the ACAT2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10937-5ug
Additional Details : Weight : 0.01000kg
Specifications
ACAT2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMNAG SDPVVIVSAA RTIIGSFNGA LAAVPVQDLG STVIKEVLKR ATVAPEDVSE VIFGHVLAAG CGQNPVRQAS VGAGIPYSVP AWSCQMICGS GLKAVCLAVQ SIGIGDSSIV VAGGMENMSK APHLAYLRTG VKIGEMPLTD SILCDGLTDA FHNCHMGITA ENVAKKWQVS REDQDKVAVL SQNRTENAQK AGHFDKEIVP VLVSTRKGLI EVKTDEFPRH GSNIEAMSKL KPYFLTDGTG TVTPANASGI NDGAAAVVLM KKSEADKRGL TPLARIVSWS QVGVEPSIMG IGPIPAIKQA VTKAGWSLED VDIFEINEAF AAVSAAIVKE LGLNPEKVNI EGGAIALGHP LGASGCRILV TLLHTLERMG RSRGVAALCI GGGMGIAMCV QR | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ACAT2 Human solution containing 20mM Tris pH 8.0, 0.1M NaCl, 1mM DTT and 20% glycerol. |