missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACTA2 Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Name | ACTA2 Protein |
| Quantity | 2 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSHMEEEDS TALVCDNGSG LCKAGFAGDD APRAVFPSIV GRPRHQGVMV GMGQKDSYVG DEAQSKRGIL TLKYPIEHGI ITNWDDMEKI WHHSFYNELR VAPEEHPTLL TEAPLNPKAN REKMTQIMFE TFNVPAMYVA IQAVLSLYAS GRTTGIVLDS GDGVTHNVPI YEGYALPHAI MRLDLAGRDL TDYLMKILTE RGYSFVTTAE REIVRDIKEK LCYVALDFEN EMATAASSSS LEKSYELPDG QVITIGNERF RCPETLFQPS FIGMESAGIH ETTYNSIMKC DIDIRKDLYA NNVLSGGTTM YPGIADRMQK EITALAPSTM KIKIIAPPER KYSVWIGGSI LASLSTFQQM WISKQEYDEA GPSIVHRKCF |
| Buffer | The ACTA2 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction