missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACTR3 Protein
A cDNA sequence encoding the ACTR3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10954-20ug
Additional Details : Weight : 0.01000kg
Specifications
ACTR3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAGRLPA CVVDCGTGYT KLGYAGNTEP QFIIPSCIAI KESAKVGDQA QRRVMKGVDD LDFFIGDEAI EKPTYATKWP IRHGIVEDWD LMERFMEQVI FKYLRAEPED HYFLLTEPPL NTPENREYTA EIMFESFNVP GLYIAVQAVL ALAASWTSRQ VGERTLTGTV IDSGDGVTHV IPVAEGYVIG SCIKHIPIAG RDITYFIQQL LRDREVGIPP EQSLETAKAV KERYSYVCPD LVKEFNKYDT DGSKWIKQYT GINAISKKEF SIDVGYERFL GPEIFFHPEF ANPDFTQPIS EVVDEVIQNC PIDVRRPLYK NIVLSGGSTM FRDFGRRLQR DLKRTVDARL KLSEELSGGR LKPKPIDVQV ITHHMQRYAV WFGGSMLAST PEFYQVCHTK KDYEEIGPSI CRHNPVFGVM S | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ACTR3 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |