missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADAM10 Protein
Click to view available options
Quantity:
1 μg
1 mg
10 μg
2 μg
5 μg
50 μg
Unit Size:
10µg
1mg
1µg
2µg
50µg
5µg
Specifications
Specifications
| Name | ADAM10 Protein |
| Quantity | 1 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MVLLRVLILL LSWAAGMGGQ YGNPLNKYIR HYEGLSYNVD SLHQKHQRAK RAVSHEDQFL RLDFHAHGRH FNLRMKRDTS LFSDEFKVET SNKVLDYDTS HIYTGHIYGE EGSFSHGSVI DGRFEGFIQT RGGTFYVEPA ERYIKDRTLP FHSVIYHEDD INYPHKYGPQ GGCADHSVFE RMRKYQMTGV EEVTQIPQEE HAANGPELLR KKRTTSAEKN TCQLYIQTDH LFFKYYGTRE AVIAQISSHV KAIDTIYQTT DFSGIRNISF MVKRIRINTT ADEKDPTNPF RFPNIGVEKF LELNSEQNHD DYCLAYVFTD RDFDDGVLGL AWVGAPSGSS GGICEKSKLY SDGKKKSLNT GIITVQNYGS HVPPKVSHIT FAHEVGHNFG SPHDSGTECT PGESKNLGQK ENGNYIMYAR ATSGDKLNNN KFSLCSIRNI SQVLEKKRNN CFVESGQPIC GNGMVEQGEE CDCGYSDQCK DECCFDANQP EGRKCKLKPG KQCSPSQGPC CTAQCAFKSK SEKCRDDSDC AREGICNGFT ALCPASDPKP NFTDCNRHTQ VCINGQCAGS ICEKYGLEEC TCASSDGKDD KELCHVCCMK KMDPSTCAST GSVQWSRHFS GRTITLQPGS PCNDFRGYCD VFMRCRLVDA DGPLARLKKA IFSPELYENI AEWIVAHWWA VLLMGIALIM LMAGFIKICS VHTPSSNPKL PPPKPLPGTL KRRRPPQPIQ QPQRQRPRES YQMGHMRR |
| Buffer | The ADAM10 solution contains 20mM Tris (pH 8), 1mM EDTA and 50% Glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction