missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKAP7 Protein
A cDNA sequence encoding the AKAP7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10993-1mg
Additional Details : Weight : 0.01000kg
Specifications
AKAP7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMGQLCC FPFSRDEGKI SEKNGGEPDD AELVRLSKRL VENAVLKAVQ QYLEETQNKN KPGEGSSVKT EAADQNGNDN ENNRK | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The AKAP7 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 50mM NaCl, 1mM DTT and 10% glycerol. |