missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ASPA Protein

A cDNA sequence encoding the ASPA was constructed and used to recombinantly synthesize the protein.

137.00€ - 5450.00€

Specifications

Name ASPA Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15946108
View Documents Promotion Details
enQuireBio™
QP11089-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15936108
View Documents Promotion Details
enQuireBio™
QP11089-20UG
20 μg
212.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15926108
View Documents Promotion Details
enQuireBio™
QP11089-1MG
1 mg
5450.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

ASPA Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The ASPA solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 1mM DTT, 0.1M NaCl and 0.1mM PMSF.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMTSCHIA EEHIQKVAIF GGTHGNELTG VFLVKHWLEN GAEIQRTGLE VKPFITNPRA VKKCTRYIDC DLNRIFDLEN LGKKMSEDLP YEVRRAQEIN HLFGPKDSED SYDIIFDLHN TTSNMGCTLI LEDSRNNFLI QMFHYIKTSL APLPCYVYLIEHPSLKYATT RSIAKYPVGI EVGPQPQGVL RADILDQMRK MIKHALDFIH HFNEGKEFPP CAIEVYKIIE KVDYPRDENG EIAAIIHPNL QDQDWKPLHP GDPMFLTLDG KTIPLGGDCT VYPVFVNEAA YYEKKEAFAK TTKLTLNAKS IRCCLH
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications