missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ATP5C1 Protein

A cDNA sequence encoding the ATP5C1 was constructed and used to recombinantly synthesize the protein.

137.00€ - 4090.00€

Specifications

Name ATP5C1 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15946168
View Documents Promotion Details
enQuireBio™
QP11107-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15936168
View Documents Promotion Details
enQuireBio™
QP11107-20UG
20 μg
212.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15926168
View Documents Promotion Details
enQuireBio™
QP11107-1MG
1 mg
4090.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 16-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

ATP5C1 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
ATP5C1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.4M Urea and 10% glycerol.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSATLKDIT RRLKSIKNIQ KITKSMKMVA AAKYARAERE LKPARIYGLG SLALYEKADI KGPEDKKKHL LIGVSSDRGL CGAIHSSIAK QMKSEVATLT AAGKEVMLVG IGDKIRGILY RTHSDQFLVA FKEVGRKPPT FGDASVIALE LLNSGYEFDE GSIIFNKFRS VISYKTEEKP IFSLNTVASA DSMSIYDDID ADVLQNYQEY NLANIIYYSL KESTTSEQSA RMTAMDNASK NASEMIDKLT LTFNRTRQAV ITKELIEIIS GAAALD
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications