missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human B7-H7 Fc Biotinylated Protein

Product Code. 18725698
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. Quantity unitSize
18725698 50 μg 50µg
1 options
This item is not returnable. View return policy

Product Code. 18725698

Brand: R&D Systems™ BT8084050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Human TMIGD2/CD28H Fc Chimera (Catalog # 8316-TR ) is immobilized at 0.125 μg/mL, 100 μL/well, the concentration of Biotinylated Recombinant Human B7-H7/HHLA2 Fc Chimera that produces 50% of the optimal binding response is 0.025-0.125 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UM44
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 11148
Molecular Weight (g/mol) M.W. (theoretical): 64 kDa (unlabeled)
Name B7H7, HERV-H LTR-associating 2, HERV-H LTR-associating protein 2, HHLA2, Human endogenous retrovirus-H long terminal repeat-associating protein 2
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human B7-H7/HHLA2 protein Human B7-H7/HHLA2 (Ile23-Asn344) (N-terminus) Accession # Q9UM44 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements 12 months from date of receipt, <= -20 °C as supplied. 2 weeks, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Biotin
Species Human
Reconstitution Reconstitute at 200 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein B7-H7/HHLA2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.