missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human B7-H7/HHLA2 Fc Chimera Protein
Click to view available options
:
50ug; Carrier Free
Description
Measured by its ability to inhibit anti-CD3 antibody induced IL-2 secretion in human T lymphocytes. The ED50 for this effect is 0.075-0.75 μg/mL.
Specifications
Specifications
| Accession Number | Q9UM44 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 11148 |
| Molecular Weight (g/mol) | M.W. (Observed): 90-100 kDa, reducing conditions; M.W. (theoretical): 64 kDa (monomer) |
| Name | B7-H7 Fc Chimera |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human B7-H7/HHLA2 protein Human B7-H7/HHLA2 (Ile23-Asn344) (N-terminus) Accession # Q9UM44 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction