missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BEND6 Protein

A cDNA sequence encoding the BEND6 was constructed and used to recombinantly synthesize the protein.

137.00€ - 4090.00€

Specifications

Name BEND6 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15926318
View Documents Promotion Details
enQuireBio™
QP11153-5UG
5 μg
137.00€
5µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15916318
View Documents Promotion Details
enQuireBio™
QP11153-20UG
20 μg
212.00€
20µg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15906318
View Documents Promotion Details
enQuireBio™
QP11153-1MG
1 mg
4090.00€
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

BEND6 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
BEND6 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMQKIVQT DEITNTQAFR KGKRKRTETM DSENANSDMD KGQRDPYSGN AFLPGESSSE DEEPLAELSK EELCAKIKSL KEKLTNTRKE NSRLRQSLVM LQVLPQAVTQ FEELVGMAEA LLKGGGTMST SASTLWRATN NSSPDSFAST CSNSNSNSSS PVSLKPEEEH QTDEKQFQIE KWQIARCNKS KPQKFINDLM QVLYTNEYMA THSLTGAKSS TSRDKAVKPA MNQNEVQEII GVTKQLFPNT DDVSIRRMIG QKLNNCTKKP NLSKNLNSQD IK
Greater than 90.0% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications