missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human bFGF / FGF2 Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
50µg
Specifications
Specifications
| Accession Number | NP_001997.5 |
| Gene ID (Entrez) | 2247 |
| Name | bFGF / FGF2 Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | FGF2 |
| Biological Activity | Shows bioactivity at a concentration of 5 ng/mL in the 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction