missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BPNT1 Protein

A cDNA sequence encoding the BPNT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11199-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15926458

  • 7200.00€ / 1mg

Estimated Shipment: 27-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

BPNT1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The BPNT1 protein solution (0.5 mg/1ml) is formulated in 20mM Tris-HCl buffer (pH 8.0) 5mM DTT, 0.1M NaCl and 10% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMASS NTVLMRLVAS AYSIAQKAGM IVRRVIAEGD LGIVEKTCAT DLQTKADRLA QMSICSSLAR KFPKLTIIGE EDLPSEEVDQ ELIEDSQWEE ILKQPCPSQY SAIKEEDLVV WVDPLDGTKE YTEGLLDNVT VLIGIAYEGK AIAGVINQPY YNYEAGPDAV LGRTIWGVLG LGAFGFQLKE VPAGKHIITT TRSHSNKLVT DCVAAMNPDA VLRVGGAGNK IIQLIEGKAS AYVFASPGCK KWDTCAPEVI LHAVGGKLTD IHGNVLQYHK DVKHMNSAGV LATLRNYDYY ASRVPESIKN ALVP
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions